5-Bromo-2-methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-(hydroxymethyl)anisole

A439263-10G A439263-1G A439263-5G
76877-362EA 60.48 USD
76877-362 76877-364 76877-366
5-Bromo-2-methoxybenzyl alcohol 98%
5-Bromo-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
MDL Number: MFCD00004630
CAS Number: 80866-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR