5-Bromo-2-chloronicotinamide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-chloropyridine-3-carboxamide
A438592-10G
A438592-1G
A438592-250MG
A438592-5G
76877-354EA
414.82
USD
76877-354
76877-356
76877-358
76877-360
5-Bromo-2-chloronicotinamide 95%
5-Bromo-2-chloronicotinamide
Precaución: for Research Use Only
Formula:
C₆H₄BrClN₂O MW: 235.4658 g/mol |
MDL Number:
MFCD13176452 CAS Number: 75291-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...