2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,2,2-Trifluoroethoxy)pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2,2,2-trifluoroethoxy)pyridine

A437999-100MG A437999-1G A437999-250MG A437999-5G
76877-346EA 39 USD
76877-346 76877-348 76877-350 76877-352
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester 98%
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₃NO₃
MDL Number: MFCD07781172
CAS Number: 1073354-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR