(4-(N,N-Diethylsulfamoyl)-2-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A435789-100MG A435789-1G A435789-250MG
76877-310EA 40 USD
76877-310 76877-312 76877-314
(4-(N,N-Diethylsulfamoyl)-2-methylphenyl)boronic acid 98%
(4-(N,N-Diethylsulfamoyl)-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈BNO₄S
MDL Number: MFCD13195674
CAS Number: 1217501-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR