4-(4-Bromo-2-fluorobenzyl)morpholine 97%

Supplier: Ambeed
Warning

A435587-1G A435587-250MG A435587-5G
76877-304EA 55.17 USD
76877-304 76877-306 76877-308
4-(4-Bromo-2-fluorobenzyl)morpholine 97%
4-(4-Bromo-2-fluorobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO
MDL Number: MFCD12022322
CAS Number: 338454-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR