4-Hydroxy-2(5H)-furanone 96%

Supplier: Ambeed
Warning

Synonyms: Tetronic acid, beta-Tetronic acid, 2,4(3H,5H)-Furandione, Tetrahydrofuran-2,4-dione

A434443-1G A434443-250MG A434443-5G
76877-294EA 44.56 USD
76877-294 76877-296 76877-298
4-Hydroxy-2(5H)-furanone 96%
4-Hydroxy-2(5H)-furanone (Tetrahydrofuran-2,4-dione, Tetronic acid, β-Oxo-γ-butyrolactone)

Precaución: for Research Use Only

Formula: C₄H₄O₃
MDL Number: MFCD00052187
CAS Number: 541-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR