2-(4-Bromo-5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A432235-1G A432235-250MG A432235-5G
76877-280EA 126.25 USD
76877-280 76877-282 76877-284
2-(4-Bromo-5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-Bromo-5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BBrClO₂S
MW: 323.44 g/mol
MDL Number: MFCD12405463
CAS Number: 942070-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR