Benazepril hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-((S)-3-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid hydrochloride, (3S)-3-[(1S)-1-Ethoxycarbonyl-3-phenylpropyl]amino-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid Hydrochloride

A430288-100MG A430288-1G A430288-250MG A430288-25G A430288-50MG A430288-5G
76877-260EA 7 USD
76877-260 76877-262 76877-264 76877-266 76877-268 76877-270
Benazepril hydrochloride 98%
Benazepril hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₅
MW: 460.96 g/mol
MDL Number: MFCD00895734
CAS Number: 86541-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR