2-(Ethoxycarbonylmethylamino)-5-pyrimidineboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-((5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)amino)acetate
A429770-100MG
A429770-1G
A429770-250MG
A429770-5G
76877-250EA
37.14
USD
76877-250
76877-252
76877-254
76877-256
2-(Ethoxycarbonylmethylamino)-5-pyrimidineboronic acid pinacol ester 96%
2-(Ethoxycarbonylmethylamino)-5-pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₂BN₃O₄ MW: 307.1500 g/mol |
MDL Number:
MFCD12546563 CAS Number: 1202805-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...