4-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A429313-1G A429313-250MG A429313-5G
76877-244EA 188.99 USD
76877-244 76877-246 76877-248
4-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%
4-(Methoxycarbonyl)indole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₄
MDL Number: MFCD11858381
CAS Number: 1072811-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR