2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(Bromomethyl)-4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A427747-100MG A427747-1G A427747-250MG A427747-5G
76877-236EA 36.08 USD
76877-236 76877-238 76877-240 76877-242
2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester 98%
2-(Bromomethyl)-4-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrClO₂
MW: 331.4409 g/mol
MDL Number: MFCD09475815
CAS Number: 957034-64-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR