3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A427472-1G A427472-5G
76877-232EA 13.8 USD
76877-232 76877-234
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄BNO₂
MW: 215.05 g/mol
MDL Number: MFCD09750437
CAS Number: 214360-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR