5-Chloro-2-cyanobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Chloro-2-cyanophenyl)boronic acid

A427023-1G A427023-250MG
76877-222EA 90 USD
76877-222 77884-101
5-Chloro-2-cyanobenzeneboronic acid 98%
5-Chloro-2-cyanobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClNO₂
MDL Number: MFCD10696673
CAS Number: 1072946-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR