4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-[2-(1-Piperidyl)ethoxy]benzoic Acid Hydrochloride

A426746-100G A426746-10G A426746-1G A426746-25G A426746-5G
76877-212EA 150 USD
76877-212 76877-214 76877-216 76877-218 76877-220
4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride 95%
4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD06657755
CAS Number: 84449-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR