3,6-Difluorosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3,6-Difluoro-2-hydroxybenzaldehyde

A425562-100MG A425562-1G A425562-250MG
76877-204EA 35 USD
76877-204 76877-206 76877-208
3,6-Difluorosalicylaldehyde 98%
3,6-Difluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MDL Number: MFCD08275281
CAS Number: 502762-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR