4-Methyl-1H-benzo[d]imidazole-2(3H)-thione 95%

Supplier: Ambeed
Warning

A424960-1G A424960-250MG
76877-196EA 116.7 USD
76877-196 76877-198
4-Methyl-1H-benzo[d]imidazole-2(3H)-thione 95%
4-Methyl-1H-benzo[d]imidazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MW: 164.22 g/mol
MDL Number: MFCD03265647
CAS Number: 27231-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR