N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester 95%

Supplier: Ambeed
Warning

A424790-100MG A424790-1G A424790-250MG
76877-190EA 89 USD
76877-190 76877-192 76877-194
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester 95%
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD09054707
CAS Number: 1239589-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR