1-Benzyl 3-methyl pyrrolidine-1,3-dicarboxylate 98%

Supplier: Ambeed
Warning

A424693-1G A424693-250MG A424693-5G
76877-184EA 50.93 USD
76877-184 76877-186 76877-188
1-Benzyl 3-methyl pyrrolidine-1,3-dicarboxylate 98%
1-Benzyl 3-methyl pyrrolidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.28 g/mol
MDL Number: MFCD06410557
CAS Number: 188847-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR