tert-Butyl 6-bromo-3,4-dihydroquinoline-1(2H)-carboxylate 97%

Supplier: Ambeed
Warning

A424289-100MG A424289-250MG
76877-176EA 11.67 USD
76877-176 76877-178
tert-Butyl 6-bromo-3,4-dihydroquinoline-1(2H)-carboxylate 97%
tert-Butyl 6-bromo-3,4-dihydroquinoline-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₂
MW: 312.20 g/mol
MDL Number: MFCD11858570
CAS Number: 1123169-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR