(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-4-trifluoromethylphenylboronic acid

A424056-100MG A424056-1G A424056-250MG
76877-170EA 37.14 USD
76877-170 76877-172 76877-174
(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid 98%
(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
MDL Number: MFCD18087684
CAS Number: 1260518-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR