4-(5-Bromothiazol-2-yl)morpholine 98%

Supplier: Ambeed
Danger

A423890-1G A423890-5G
76877-166EA 123.07 USD
76877-166 76877-168
4-(5-Bromothiazol-2-yl)morpholine 98%
4-(5-Bromothiazol-2-yl)morpholine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂OS
MW: 249.128 g/mol
MDL Number: MFCD09879069
CAS Number: 933728-73-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR