N-Boc-D-alanine methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Ala-OMe, BOC-D-Alanine methyl ester, (R)-Methyl 2-((tert-butoxycarbonyl)amino)propanoate, BOC-D-Alanine methyl este

A422757-100G A422757-10G A422757-1G A422757-25G A422757-500G A422757-5G
76877-148EA 187.78 USD
76877-148 76877-150 76877-152 76877-154 76877-156 76877-158
N-Boc-D-alanine methyl ester 97%
N-Boc-D-alanine methyl ester

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD00191865
CAS Number: 91103-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR