3-Bromo-2,4-dichloropyridine 97%

Supplier: Ambeed
Warning

A421692-1G A421692-250MG A421692-25G A421692-5G
76877-134EA 11.67 USD
76877-134 76877-136 76877-138 76877-140
3-Bromo-2,4-dichloropyridine 97%
3-Bromo-2,4-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MW: 226.886 g/mol
MDL Number: MFCD14705096
CAS Number: 144584-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR