4'-Trifluoromethoxyacetophenone 98%

Supplier: Ambeed
Warning

A421321-100G A421321-10G A421321-1G A421321-25G A421321-500G A421321-5G
76877-114EA 59 USD
76877-114 76877-116 76877-118 76877-120 76877-122 76877-124
4'-Trifluoromethoxyacetophenone 98%
4'-Trifluoromethoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD00042404
CAS Number: 85013-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR