7-(Trifluoromethyl)-1H-indole 97%

Supplier: Ambeed
Warning

A421060-100MG A421060-1G A421060-250MG A421060-5G
76877-106EA 60.48 USD
76877-106 76877-108 76877-110 76877-112
7-(Trifluoromethyl)-1H-indole 97%
7-(Trifluoromethyl)-1H-indole

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
MDL Number: MFCD09954769
CAS Number: 172217-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR