5-Bromo-2-(dimethylamino)benzaldehyde 98%

Supplier: Ambeed
Warning

A418940-1G A418940-5G
76877-092EA 33 USD
76877-092 76877-094
5-Bromo-2-(dimethylamino)benzaldehyde 98%
5-Bromo-2-(dimethylamino)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MDL Number: MFCD20529529
CAS Number: 171881-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR