2-Bromo-N1,N1-dimethylbenzene-1,4-diamine 98%

Supplier: Ambeed
Warning

A418901-100MG A418901-1G A418901-250MG A418901-5G
76877-084EA 87 USD
76877-084 76877-086 76877-088 76877-090
2-Bromo-N1,N1-dimethylbenzene-1,4-diamine 98%
2-Bromo-N1,N1-dimethylbenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂
MW: 215.09 g/mol
MDL Number: MFCD09966038
CAS Number: 860444-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR