N,N,N',N'-Tetrakis(p-tolyl)benzidine 98%

Supplier: Ambeed
Warning

A418014-100G A418014-25G A418014-500G A418014-5G
76877-062EA 98 USD
76877-062 76877-064 76877-066 76877-068
N,N,N',N'-Tetrakis(p-tolyl)benzidine 98%
N,N,N',N'-Tetrakis(p-tolyl)benzidine

Precaución: for Research Use Only

Formula: C₄₀H₃₆N₂
MDL Number: MFCD00799300
CAS Number: 76185-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR