Bis(2-bromoethyl)ether 98%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether

A416970-100G A416970-25G A416970-5G
76877-050EA 54 USD
76877-050 76877-052 76877-054
Bis(2-bromoethyl)ether 98%
Bis(2-bromoethyl)ether

Precaución: for Research Use Only

Formula: C₄H₈Br₂O
MDL Number: MFCD00039196
CAS Number: 5414-19-7
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR