2-(Bromomethyl)-2-butylhexanoic acid 97%

Supplier: Ambeed
Warning

A415929-10G A415929-25G A415929-1G A415929-5G
76877-044EA 160 USD
76877-044 76877-046 77884-077 77884-078
2-(Bromomethyl)-2-butylhexanoic acid 97%
2-(Bromomethyl)-2-butylhexanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrO₂
MDL Number: MFCD18206921
CAS Number: 100048-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR