4-Carbamoyl-3-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carbamoyl-3-fluorophenylboronic acid, (4-Carbamoyl-3-fluorophenyl)boronic acid

A414874-1G A414874-250MG A414874-5G
76877-028EA 157.02 USD
76877-028 76877-030 76877-032
4-Carbamoyl-3-fluorobenzeneboronic acid 98%
4-Carbamoyl-3-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
MDL Number: MFCD09258667
CAS Number: 874288-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR