(R)-2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole 98%
Supplier: Ambeed
|
Warning
|
A414150-25G
A414150-5G
A414150-1G
A414150-250MG
A414150-10MG
76877-024EA
330
USD
76877-024
76877-026
77673-010
77673-012
77884-072
(R)-2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole 98%
(R)-2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole
Precaución: for Research Use Only
Formula:
C₁₆H₁₄F₃N₃O₂S |
MDL Number:
MFCD13196699 CAS Number: 138530-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...