5-Bromo-1-chloro-2,3-difluorobenzene 98%

Supplier: Ambeed
Warning

A413267-1G A413267-250MG A413267-5G A413267-25G
76877-012EA 33 USD
76877-012 76877-014 76877-016 77884-068
5-Bromo-1-chloro-2,3-difluorobenzene 98%
5-Bromo-1-chloro-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD11845915
CAS Number: 1060813-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR