Tipifarnib 98%

Supplier: Ambeed
Warning

A412936-100MG A412936-10MG A412936-1MG A412936-2MG A412936-50MG A412936-5MG
76877-000EA 452 USD
76877-000 76877-002 76877-004 76877-006 76877-008 76877-010
Tipifarnib 98%
Tipifarnib

Precaución: for Research Use Only

Formula: C₂₇H₂₂Cl₂N₄O
MDL Number: MFCD07772347
CAS Number: 192185-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR