2-Ethylphenol 99.9%

Supplier: Ambeed
Danger

Synonyms: 1-Ethyl-2-hydroxybenzene

A412005-25G A412005-5G
76876-996EA 12.74 USD
76876-996 76876-998
2-Ethylphenol 99.9%
2-Ethylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00002249
CAS Number: 90-00-6
UN: 3145
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR