2-Methyl-5-(-1-pyrrolidinylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

A411058-100MG A411058-1G A411058-250MG A411058-5G
76876-984EA 39 USD
76876-984 76876-986 76876-988 76876-990
2-Methyl-5-(-1-pyrrolidinylsulfonyl)benzeneboronic acid 98%
2-Methyl-5-(-1-pyrrolidinylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄S
MDL Number: MFCD08056365
CAS Number: 871333-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR