5-Bromo-1,3-difluoro-2-(2,2,2-trifluoroethoxy)benzene 97%

Supplier: Ambeed
Warning

A409726-100MG A409726-1G A409726-250MG A409726-5G
76876-972EA 22.28 USD
76876-972 76876-974 76876-976 76876-978
5-Bromo-1,3-difluoro-2-(2,2,2-trifluoroethoxy)benzene 97%
5-Bromo-1,3-difluoro-2-(2,2,2-trifluoroethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₄BrF₅O
MW: 291.01 g/mol
MDL Number: MFCD19237199
CAS Number: 145767-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR