1,3-Diphenyltetramethyldisilazane 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetramethyl-1,3-diphenyldisilazane, Bis(dimethyl(phenyl)silyl)amine
A406986-5G
A406986-1G
76876-948EA
44
USD
76876-948
77613-248
1,3-Diphenyltetramethyldisilazane 95%
1,3-Diphenyltetramethyldisilazane
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NSi₂ |
MDL Number:
MFCD00042916 CAS Number: 3449-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...