3-Carboxy-5-fluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-fluorobenzeneboronic acid pinacol ester

A405683-1G A405683-250MG A405683-5G
76876-940EA 76.39 USD
76876-940 76876-942 76876-944
3-Carboxy-5-fluorophenylboronic acid pinacol ester 98%
3-Carboxy-5-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
MW: 266.07 g/mol
MDL Number: MFCD12756465
CAS Number: 936728-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR