N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide 98%

Supplier: Ambeed
Warning

A405553-100MG A405553-1G A405553-250MG
76876-930EA 39 USD
76876-930 76876-932 76876-934
N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide 98%
N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₃
MDL Number: MFCD10700144
CAS Number: 1218790-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR