Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylic acid Benzyl ester, N-Cbz-7-oxa-3-azabicyclo[4.1.0]heptane

A405333-1G A405333-5G
76876-926EA 48.81 USD
76876-926 76876-928
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 95%
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
MDL Number: MFCD12407051
CAS Number: 66207-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR