2-(3-Methoxy-4-(trifluoromethyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

A401905-100MG A401905-1G A401905-250MG
76876-902EA 38.2 USD
76876-902 76876-904 76876-906
2-(3-Methoxy-4-(trifluoromethyl)phenyl)acetic acid 95%
2-(3-Methoxy-4-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
MDL Number: MFCD13194423
CAS Number: 1214372-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR