2-Amino-4-chloronicotinaldehyde 97%

Supplier: Ambeed
Warning

A401398-250MG A401398-5G A401398-1G
76876-900EA 49 USD
76876-900 77551-204 77594-218
2-Amino-4-chloronicotinaldehyde 97%
2-Amino-4-chloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MDL Number: MFCD17016043
CAS Number: 884004-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR