4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-[4-(3-Bromopropoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-(3-Bromopropoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A401296-100MG A401296-1G A401296-250MG A401296-5G
76876-892EA 12 USD
76876-892 76876-894 76876-896 76876-898
4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester 97%
4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BBrO₃
MDL Number: MFCD09258657
CAS Number: 957061-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR