4-Chloro-1-iodo-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A401009-100G A401009-1G A401009-250MG A401009-25G A401009-5G A401009-10G
76876-882EA 183 USD
76876-882 76876-884 76876-886 76876-888 76876-890 77884-049
4-Chloro-1-iodo-2-nitrobenzene 97%
4-Chloro-1-iodo-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MDL Number: MFCD11226294
CAS Number: 5446-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR