(±)-2-Phenylbutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: (±)-2-Phenylbutanoic acid, 2-Phenylbutanoic acid

A398361-500G A398361-100G A398361-25G
76876-874EA 180.36 USD
76876-874 77551-192 77572-228
(±)-2-Phenylbutyric acid 98%
(±)-2-Phenylbutyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00002667
CAS Number: 90-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR