cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (cis-3-carboxycyclopent-1-yl)carbamate, cis-3-(Boc-amino)cyclopentanecarboxylic acid, cis-3-[(tert-Butoxycarbonyl)amino]cyclopentane-1-carboxylic acid

A397314-100MG A397314-1G A397314-250MG A397314-5G
76876-866EA 7.43 USD
76876-866 76876-868 76876-870 76876-872
cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected 95%
cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.273 g/mol
MDL Number: MFCD09992879
CAS Number: 410090-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR