tert-Butyl (5-chlorothiophen-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A397105-100MG A397105-1G A397105-250MG A397105-5G
76876-858EA 11 USD
76876-858 76876-860 76876-862 76876-864
tert-Butyl (5-chlorothiophen-2-yl)carbamate 98%
tert-Butyl (5-chlorothiophen-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂S
MDL Number: MFCD06659528
CAS Number: 63806-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR