3'-Chloro-[1,1'-biphenyl]-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A396776-1G A396776-250MG A396776-5G
76876-852EA 101.85 USD
76876-852 76876-854 76876-856
3'-Chloro-[1,1'-biphenyl]-4-amine hydrochloride 97%
3'-Chloro-[1,1'-biphenyl]-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₁Cl₂N
MW: 240.12 g/mol
MDL Number: MFCD06739433
CAS Number: 811842-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR