5-Fluoro-2-methoxy-3-pyridineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A395986-100MG A395986-1G A395986-250MG A395986-5G
76876-846EA 37 USD
76876-846 76876-848 76876-850 77884-039
5-Fluoro-2-methoxy-3-pyridineboronic acid pinacol ester 95%
5-Fluoro-2-methoxy-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₃
MDL Number: MFCD08063053
CAS Number: 1083168-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR